Product Details

SNP ID
rs9955128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63755488 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTAGTTTGTTTCAGTAGCAGACA[C/T]TGAAGTTTGAGGTTTAGCTGAGCAA
Phenotype
MIM: 603357
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINB7 PubMed Links

Gene Details

Gene
SERPINB7
Gene Name
serpin family B member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040147.2 Intron NP_001035237.1
NM_001261830.1 Intron NP_001248759.1
NM_001261831.1 Intron NP_001248760.1
NM_003784.3 Intron NP_003775.1

View Full Product Details