Product Details

SNP ID
rs10187155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:28827452 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTACTGGAAATAGTTATTCTTAC[G/T]TTCTGTGTCTGTTCCATTACTTTCA
Phenotype
MIM: 614029
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SPDYA PubMed Links

Gene Details

Gene
SPDYA
Gene Name
speedy/RINGO cell cycle regulator family member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008779.1 Intron NP_001008779.1
NM_001142634.1 Intron NP_001136106.1
NM_182756.3 Intron NP_877433.2

View Full Product Details