Product Details

SNP ID
rs9410439
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:89009139 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTCTTTTCTAAGTTGATCCCCAG[C/G]CCCTTGCATGGCTCCAGTTTCCTCT
Phenotype
MIM: 601965 MIM: 605263
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
S1PR3 PubMed Links

Gene Details

Gene
S1PR3
Gene Name
sphingosine-1-phosphate receptor 3
There are no transcripts associated with this gene.

Gene
SHC3
Gene Name
SHC adaptor protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016848.5 6400 UTR 3 NP_058544.3
XM_011518785.2 6400 UTR 3 XP_011517087.1
XM_011518786.2 6400 UTR 3 XP_011517088.1
XM_017014809.1 6400 Intron XP_016870298.1
XM_017014810.1 6400 Intron XP_016870299.1

View Full Product Details