Product Details

SNP ID
rs7539459
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:150067559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCGCTGGGCCTCCAGCACCAAAG[G/T]GCAATAGGGGTCTGAGAGTCCGGGC
Phenotype
MIM: 610035
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
VPS45 PubMed Links

Gene Details

Gene
VPS45
Gene Name
vacuolar protein sorting 45 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001279353.1 267 UTR 5 NP_001266282.1
NM_001279354.1 267 Intron NP_001266283.1
NM_001279355.1 267 Intron NP_001266284.1
NM_007259.4 267 UTR 5 NP_009190.2
XM_017000164.1 267 Intron XP_016855653.1

View Full Product Details