Product Details

SNP ID
rs6634518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:143627809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTCACGTAACTGAGAACAAACT[C/T]TAGTGTTAGTTTAATTTGAGCCATG
Phenotype
MIM: 300562
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLITRK4 PubMed Links

Gene Details

Gene
SLITRK4
Gene Name
SLIT and NTRK like family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184749.2 3526 UTR 3 NP_001171678.1
NM_001184750.2 3526 UTR 3 NP_001171679.1
NM_173078.4 3526 UTR 3 NP_775101.1
XM_005262365.4 3526 Intron XP_005262422.1
XM_011531265.2 3526 Intron XP_011529567.1

View Full Product Details