Product Details

SNP ID
rs9461901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33698940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTTTACCAGCCTAAATTTTCAC[A/G]TTAACAAAACTACTGGAGAAAAAAA
Phenotype
MIM: 147267 MIM: 614461
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITPR3 PubMed Links

Gene Details

Gene
ITPR3
Gene Name
inositol 1,4,5-trisphosphate receptor type 3
There are no transcripts associated with this gene.

Gene
MIR3934
Gene Name
microRNA 3934
There are no transcripts associated with this gene.

Gene
UQCC2
Gene Name
ubiquinol-cytochrome c reductase complex assembly factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032340.3 Intron NP_115716.1

View Full Product Details