Product Details
- SNP ID
-
rs6049715
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:24473249 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGTATCAGTGGAAAGTTGCATAA[A/G]GTCATTTTGGGGCAGTGCACTGATA
- Phenotype
-
MIM: 614311
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105372585
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116761928] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105372585
- Gene Name
- translation initiation factor IF-2-like
There are no transcripts associated with this gene.
- Gene
- SYNDIG1
- Gene Name
- synapse differentiation inducing 1
View Full Product Details