Product Details

SNP ID
rs9683949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2742753 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAGTAGGCCCTGGCGTGGTCCAC[C/T]CCCCAGCCCCTTGGTGGAAGTCCTG
Phenotype
MIM: 610669
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNIP2 PubMed Links

Gene Details

Gene
TNIP2
Gene Name
TNFAIP3 interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161527.1 Intron NP_001154999.1
NM_001292016.1 Intron NP_001278945.1
NM_024309.3 Intron NP_077285.3

View Full Product Details