Product Details

SNP ID
rs9912503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50346170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGAGCGCGCGAGTGCAGAAGCT[G/T]GTGCGGCGCTACAAGCTGGCGATTG
Phenotype
MIM: 608125
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
XYLT2 PubMed Links

Gene Details

Gene
XYLT2
Gene Name
xylosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022167.3 139 Silent Mutation CTG,CTT L10L NP_071450.2
XM_005257572.4 139 Intron XP_005257629.1

View Full Product Details