Product Details
- SNP ID
-
rs9971147
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:102138654 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTGGAGGACCAGAATGAAGTGTC[C/G]CTGCTCACGGCTCTGACGGAGATCT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PPRC1
PubMed Links
Gene Details
- Gene
- PPRC1
- Gene Name
- peroxisome proliferator-activated receptor gamma, coactivator-related 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001288727.1 |
411 |
Silent Mutation |
TCC,TCG |
S126S |
NP_001275656.1 |
NM_001288728.1 |
411 |
Intron |
|
|
NP_001275657.1 |
NM_015062.4 |
411 |
Silent Mutation |
TCC,TCG |
S126S |
NP_055877.3 |
XM_005269656.3 |
411 |
Silent Mutation |
TCC,TCG |
S126S |
XP_005269713.1 |
XM_006717731.3 |
411 |
Silent Mutation |
TCC,TCG |
S126S |
XP_006717794.1 |
XM_011539549.2 |
411 |
Silent Mutation |
TCC,TCG |
S126S |
XP_011537851.1 |
XM_011539551.2 |
411 |
Intron |
|
|
XP_011537853.1 |
XM_011539552.2 |
411 |
Silent Mutation |
TCC,TCG |
S126S |
XP_011537854.1 |
XM_017015982.1 |
411 |
Silent Mutation |
TCC,TCG |
S126S |
XP_016871471.1 |
XM_017015983.1 |
411 |
Silent Mutation |
TCC,TCG |
S126S |
XP_016871472.1 |
XM_017015984.1 |
411 |
Silent Mutation |
TCC,TCG |
S126S |
XP_016871473.1 |
XM_017015985.1 |
411 |
Silent Mutation |
TCC,TCG |
S126S |
XP_016871474.1 |
View Full Product Details