Product Details

SNP ID
rs10502045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:105040443 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAACCACCCAAGTTGGATGGGGAA[A/T]TGTAGACAGAGTGGATATAATTTAT
Phenotype
MIM: 615680 MIM: 147678
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CARD16 PubMed Links

Gene Details

Gene
CARD16
Gene Name
caspase recruitment domain family member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017534.1 1770 Intron NP_001017534.1
NM_052889.2 1770 Intron NP_443121.1
XM_011542583.2 1770 UTR 3 XP_011540885.1
Gene
CASP1
Gene Name
caspase 1
There are no transcripts associated with this gene.

View Full Product Details