Product Details

SNP ID
rs9462985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:44388555 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGCCAAAGGGCAGTAGAGTGGGA[A/G]TGAAATAGAAATTAGATTTTAGTGC
Phenotype
MIM: 602868
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC5L PubMed Links

Gene Details

Gene
CDC5L
Gene Name
cell division cycle 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253.3 Intron NP_001244.1

View Full Product Details