Product Details

SNP ID
rs10161823
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:39011217 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTTCACACTTTAAAATGCAACCA[C/G]AATTTTCAAATTGTTGTCACAATTT
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
PROSER1 PubMed Links

Gene Details

Gene
PROSER1
Gene Name
proline and serine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025138.4 3790 UTR 3 NP_079414.3
XM_005266545.3 3790 UTR 3 XP_005266602.1
XM_011535239.2 3790 UTR 3 XP_011533541.1

View Full Product Details