Product Details

SNP ID
rs10518184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:78062928 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAGCTGCATTCTGTGCCTTGAAA[C/T]TGGCATTGTTCATAGTTAGAGAACT
Phenotype
MIM: 607830
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRAS1 PubMed Links

Gene Details

Gene
FRAS1
Gene Name
Fraser extracellular matrix complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166133.1 Intron NP_001159605.1
NM_025074.6 Intron NP_079350.5
XM_006714316.2 Intron XP_006714379.1

View Full Product Details