Product Details

SNP ID
rs10157591
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:153114067 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGGCAGGTCACCTCTCCTCCCTG[G/T]GATTATAAGCAGGGGAGAATCATAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SPRR2F PubMed Links

Gene Details

Gene
SPRR2F
Gene Name
small proline rich protein 2F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014450.2 Intron NP_001014450.1
XM_017002176.1 Intron XP_016857665.1

View Full Product Details