Product Details

SNP ID
rs5962805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:107215262 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTGAGCCAATCACTGTCCATGGA[A/G]AGTATACCAATTTTTGTCAAGAATG
Phenotype
MIM: 300933
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUP62CL PubMed Links

Gene Details

Gene
NUP62CL
Gene Name
nucleoporin 62 C-terminal like
There are no transcripts associated with this gene.

Gene
PIH1D3
Gene Name
PIH1 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001169154.1 Intron NP_001162625.1
NM_173494.1 Intron NP_775765.1
XM_011530855.2 Intron XP_011529157.1

View Full Product Details