Product Details

SNP ID
rs6004917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20921571 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAGAAAGCATGAAGCTAAGACCT[A/G]AGGGTAAAGAAGAACCCAGCCTAGG
Phenotype
MIM: 602007
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRKL PubMed Links

Gene Details

Gene
CRKL
Gene Name
CRK like proto-oncogene, adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005207.3 Intron NP_005198.1

View Full Product Details