Product Details

SNP ID
rs6138304
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:24474673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTGGGGTGGCATTTAGAGAATGA[C/G]TCTGAAGGAGAGAGTACTTGCGTGG
Phenotype
MIM: 614311
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC105372585 PubMed Links

Gene Details

Gene
LOC105372585
Gene Name
translation initiation factor IF-2-like
There are no transcripts associated with this gene.

Gene
SYNDIG1
Gene Name
synapse differentiation inducing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323606.1 Intron NP_001310535.1
NM_001323607.1 Intron NP_001310536.1
NM_024893.2 Intron NP_079169.1
XM_011529347.2 Intron XP_011527649.1
XM_011529348.2 Intron XP_011527650.1
XM_011529349.2 Intron XP_011527651.1
XM_011529350.1 Intron XP_011527652.1
XM_011529351.1 Intron XP_011527653.1
XM_011529352.2 Intron XP_011527654.1
XM_011529353.2 Intron XP_011527655.1
XM_011529354.2 Intron XP_011527656.1
XM_011529356.2 Intron XP_011527658.1
XM_011529358.2 Intron XP_011527660.1
XM_017028064.1 Intron XP_016883553.1
XM_017028065.1 Intron XP_016883554.1
XM_017028066.1 Intron XP_016883555.1
XM_017028067.1 Intron XP_016883556.1
XM_017028068.1 Intron XP_016883557.1
XM_017028069.1 Intron XP_016883558.1

View Full Product Details