Product Details

SNP ID
rs9478362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:150019390 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAACCCCAGCAGCAGCTCAGATGC[C/T]CCTTCCTCTGAGTCCACTCTGCTCC
Phenotype
MIM: 611047
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAET1L PubMed Links

Gene Details

Gene
RAET1L
Gene Name
retinoic acid early transcript 1L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130900.2 Intron NP_570970.2
XM_011535486.2 Intron XP_011533788.1

View Full Product Details