Product Details

SNP ID
rs6107581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:5134390 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTCTGAGAGGGGCTGGTGTTATC[A/C]GTGTTCATAAGGATATTATTCAGAA
Phenotype
MIM: 603549 MIM: 176740
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDS2 PubMed Links

Gene Details

Gene
CDS2
Gene Name
CDP-diacylglycerol synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003818.3 Intron NP_003809.1
XM_006723660.1 Intron XP_006723723.1
XM_011529390.1 Intron XP_011527692.1
Gene
PCNA
Gene Name
proliferating cell nuclear antigen
There are no transcripts associated with this gene.

View Full Product Details