Product Details
- SNP ID
-
rs10137035
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:102778503 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCAAGAAACCTTCAGATTACGAGC[C/T]GTTGACGGGACCAGTTGTATTTCAC
- Phenotype
-
MIM: 601896
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TRAF3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs118040404] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TRAF3
- Gene Name
- TNF receptor associated factor 3
View Full Product Details