Product Details
- SNP ID
-
rs10454145
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:74498677 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCAGGCTATAAGTTGAGGGTGACAT[C/T]AGAGGCGAGTAAGAGTCAGGGCAGC
- Phenotype
-
MIM: 607164
MIM: 610231
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LBX2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79539875] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LBX2
- Gene Name
- ladybird homeobox 2
- Gene
- LBX2-AS1
- Gene Name
- LBX2 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- PCGF1
- Gene Name
- polycomb group ring finger 1
There are no transcripts associated with this gene.
- Gene
- TTC31
- Gene Name
- tetratricopeptide repeat domain 31
There are no transcripts associated with this gene.
View Full Product Details