Product Details

SNP ID
rs10106906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:28768070 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGACACAGTTAATGGCCTCTCAT[C/G]CCACTCCTCAGGTGGCTTGTGAGGG
Phenotype
MIM: 611352
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
INTS9 PubMed Links

Gene Details

Gene
INTS9
Gene Name
integrator complex subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145159.2 2118 UTR 3 NP_001138631.1
NM_001172562.1 2118 UTR 3 NP_001166033.1
NM_018250.3 2118 UTR 3 NP_060720.2
XM_006716356.2 2118 UTR 3 XP_006716419.1
XM_006716357.3 2118 Intron XP_006716420.1
XM_011544573.1 2118 UTR 3 XP_011542875.1
XM_011544574.1 2118 UTR 3 XP_011542876.1
XM_011544575.2 2118 UTR 3 XP_011542877.1
XM_011544576.1 2118 UTR 3 XP_011542878.1

View Full Product Details