Product Details

SNP ID
rs10135651
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:106491163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGCATCCTCTCCTGGCTAAGAT[G/T]TCAATAGGTCCAGTTGTGCACATTA
Phenotype
MIM: 146910 MIM: 147070 MIM: 147010
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
IGH PubMed Links

Gene Details

Gene
IGH
Gene Name
immunoglobulin heavy locus
There are no transcripts associated with this gene.

Gene
LINC00221
Gene Name
long intergenic non-protein coding RNA 221
There are no transcripts associated with this gene.

View Full Product Details