Product Details

SNP ID
rs10185075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:29062199 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGACTTGGAATTCCAGTAGCCTA[A/G]GGTTACTGGGCAACACAATGGTGTC
Phenotype
MIM: 613425
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf71 PubMed Links

Gene Details

Gene
C2orf71
Gene Name
chromosome 2 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029883.2 6537 UTR 3 NP_001025054.1
Gene
FAM179A
Gene Name
family with sequence similarity 179 member A
There are no transcripts associated with this gene.

View Full Product Details