Product Details

SNP ID
rs9369350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42211353 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGGATAACATATGACAAACTTAA[A/G]CTCTACCCACCAAGAGTTTCTATCC
Phenotype
MIM: 611976
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS10 PubMed Links

Gene Details

Gene
MRPS10
Gene Name
mitochondrial ribosomal protein S10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018141.3 Intron NP_060611.2
XM_005249206.2 Intron XP_005249263.1
XM_011514724.2 Intron XP_011513026.1
XM_011514725.2 Intron XP_011513027.1
XM_017010996.1 Intron XP_016866485.1

View Full Product Details