Product Details
- SNP ID
-
hCV29867496
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:145727590 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTACATACAATGTAAACTTTCCTTG[T/A]GAAAAAAAAGCCTCATAAAAAAAGT
- Phenotype
-
MIM: 604463
- Polymorphism
- T/A, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CD160
PubMed Links
Gene Details
- Gene
- CD160
- Gene Name
- CD160 molecule
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007053.3 |
|
Intron |
|
|
NP_008984.1 |
View Full Product Details