Product Details

SNP ID
rs9366814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33113022 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGCTGGTGTGTCCAGACTACAAG[G/T]ATCACTGAGGATTCAGTGCTCACAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HLA-DPB2 PubMed Links

Gene Details

Gene
HLA-DPB2
Gene Name
major histocompatibility complex, class II, DP beta 2 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details