Product Details

SNP ID
rs7540224
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:27838893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAATTCACATGTTACCTTTAGGCA[A/G]TATGAAAATACTCTTTGGGTTCTTT
Phenotype
MIM: 602636
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP1R8 PubMed Links

Gene Details

Gene
PPP1R8
Gene Name
protein phosphatase 1 regulatory subunit 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002713.3 Intron NP_002704.1
NM_014110.4 Intron NP_054829.2
NM_138558.2 Intron NP_612568.1
XM_017001594.1 Intron XP_016857083.1
Gene
SCARNA1
Gene Name
small Cajal body-specific RNA 1
There are no transcripts associated with this gene.

View Full Product Details