Product Details

SNP ID
rs10119273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129739627 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTGCTGGTCACAGGTGGCGGGCC[A/G]CTTCCCAGAGGATCTGCAGAGCCAT
Phenotype
MIM: 605172
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTGES PubMed Links

Gene Details

Gene
PTGES
Gene Name
prostaglandin E synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004878.4 857 Missense Mutation GCG,GTG A148V NP_004869.1
XM_011519210.2 857 Missense Mutation GCG,GTG A176V XP_011517512.1

View Full Product Details