Product Details
- SNP ID
-
rs10452329
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:722459 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGAGCTGGGTGGAGAGGAAGGCATA[C/T]GTCACACTGAGGCATCAGAGACACA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PCGF3
PubMed Links
Gene Details
- Gene
- PCGF3
- Gene Name
- polycomb group ring finger 3
View Full Product Details