Product Details

SNP ID
rs7543562
Assay Type
Functionally tested
NCBI dbSNP Submissions
55
Location
Chr.1:236143525 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTGAGCGTTGGCAGGGTGAGACC[A/G]TGTTGGTTTTGATGAGATGTTTTGA
Phenotype
MIM: 604658
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR137B PubMed Links

Gene Details

Gene
GPR137B
Gene Name
G protein-coupled receptor 137B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003272.3 Intron NP_003263.1
XM_017002209.1 Intron XP_016857698.1
XM_017002210.1 Intron XP_016857699.1

View Full Product Details