Product Details

SNP ID
rs6059153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33201562 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTGAAAAATCCTCAACTCCTCCA[A/T]GTCTCCATCGAGAGCACCCCTCAAA
Phenotype
MIM: 607627
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BPIFA4P PubMed Links

Gene Details

Gene
BPIFA4P
Gene Name
BPI fold containing family A member 4, pseudogene
There are no transcripts associated with this gene.

View Full Product Details