Product Details

SNP ID
rs10227452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92533629 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTTTAGTTAAGCTCTCTTTCAT[C/T]TGCAGCATTCTAGCCTCTACAAGAT
Phenotype
MIM: 602136
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PEX1 PubMed Links

Gene Details

Gene
PEX1
Gene Name
peroxisomal biogenesis factor 1
There are no transcripts associated with this gene.

Gene
RBM48
Gene Name
RNA binding motif protein 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032120.3 Intron NP_115496.2
XM_005250636.4 Intron XP_005250693.1
XM_006716149.3 Intron XP_006716212.1
XM_006716150.3 Intron XP_006716213.1
XM_017012709.1 Intron XP_016868198.1

View Full Product Details