Product Details

SNP ID
rs10238918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:31337755 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATAGCTTTTTATATTGTTGCAAAT[G/T]CATCTCCCAATATCATTGTCAGCTT
Phenotype
MIM: 611513
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NEUROD6 PubMed Links

Gene Details

Gene
NEUROD6
Gene Name
neuronal differentiation 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022728.3 1867 UTR 3 NP_073565.2

View Full Product Details