Product Details
- SNP ID
-
rs10193669
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:189442861 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGAAGCCAAAAAAAAATGAGTCCT[G/T]TGGCTTTGTTGACATTGATCTTATG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
WDR75
PubMed Links
Gene Details
- Gene
- WDR75
- Gene Name
- WD repeat domain 75
View Full Product Details