Product Details

SNP ID
rs6646308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:118980125 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACATTCTGCAAGCTATAAAGCTT[C/T]CTGGATGTAGTGCTCTGTCTCTACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LONRF3 PubMed Links

Gene Details

Gene
LONRF3
Gene Name
LON peptidase N-terminal domain and ring finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031855.2 Intron NP_001027026.1
NM_001289109.1 Intron NP_001276038.1
NM_024778.5 Intron NP_079054.3
XM_005262476.2 Intron XP_005262533.1
XM_011531398.2 Intron XP_011529700.1
XM_017029845.1 Intron XP_016885334.1

View Full Product Details