Product Details

SNP ID
rs9935001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:21598965 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTTCTCAGGCTTGGGGAACGGGG[A/C]AGGCGGGTCGTGGTGGTGGGGAGAG
Phenotype
MIM: 609388
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927814 PubMed Links

Gene Details

Gene
LOC101927814
Gene Name
uncharacterized LOC101927814
There are no transcripts associated with this gene.

Gene
METTL9
Gene Name
methyltransferase like 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077180.2 Intron NP_001070648.1
NM_001288659.1 Intron NP_001275588.1
NM_001288660.1 Intron NP_001275589.1
NM_016025.4 Intron NP_057109.3
XM_017023269.1 Intron XP_016878758.1

View Full Product Details