Product Details

SNP ID
rs6007761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46274497 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACATCATCCCCCCGCTGTTCCTA[C/T]GCTGGTTCCCTCATTCTCCGGGACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTC38 PubMed Links

Gene Details

Gene
TTC38
Gene Name
tetratricopeptide repeat domain 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017931.2 Intron NP_060401.2
XM_011530259.1 Intron XP_011528561.1
XM_011530260.2 Intron XP_011528562.1

View Full Product Details