Product Details

SNP ID
rs10160562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125836107 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAACCCAGATTATTCTTATGCCC[A/G]AAAGTGATGGCAAGACTTTTTTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PATE4 PubMed Links

Gene Details

Gene
PATE4
Gene Name
prostate and testis expressed 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144874.1 Intron NP_001138346.1

View Full Product Details