Product Details

SNP ID
rs6015228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58238940 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTCTTCCCCTGGGTGACTTCTGG[A/G]CAAGTCAGGTCACTACCTGGGGGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP4R1L PubMed Links

Gene Details

Gene
PPP4R1L
Gene Name
protein phosphatase 4 regulatory subunit 1 like (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details