Product Details

SNP ID
rs5760192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20721371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGATGAACTGGTGTGCCAGAAG[C/T]TGGGATTTAGACGCTGCCCACAGAA
Phenotype
MIM: 600286
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PI4KA PubMed Links

Gene Details

Gene
PI4KA
Gene Name
phosphatidylinositol 4-kinase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058004.3 5130 Silent Mutation CAA,CAG Q1681Q NP_477352.3

View Full Product Details