Product Details
- SNP ID
-
rs9478099
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:169730151 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAAATGAAATTTGCTTGAATATTCT[C/G]AAAGTGAAAATTGCACAGTGCATAC
- Phenotype
-
MIM: 613069
MIM: 186977
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PHF10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77356976] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PHF10
- Gene Name
- PHD finger protein 10
There are no transcripts associated with this gene.
- Gene
- TCTE3
- Gene Name
- t-complex-associated-testis-expressed 3
View Full Product Details