Product Details

SNP ID
rs6622896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:84058431 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATCTTGAGAACTGTTGCTCACAGA[A/G]TTTTAACAAGAACAAAAATAAATAT
Phenotype
MIM: 300303
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPS6KA6 PubMed Links

Gene Details

Gene
RPS6KA6
Gene Name
ribosomal protein S6 kinase A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014496.4 8120 UTR 3 NP_055311.1
XM_011530917.2 8120 Intron XP_011529219.1
XM_011530918.2 8120 Intron XP_011529220.1
XM_011530919.2 8120 Intron XP_011529221.1
XM_011530920.2 8120 Intron XP_011529222.1
XM_017029423.1 8120 Intron XP_016884912.1
XM_017029424.1 8120 Intron XP_016884913.1
XM_017029425.1 8120 Intron XP_016884914.1
XM_017029426.1 8120 Intron XP_016884915.1

View Full Product Details