Product Details

SNP ID
rs7523420
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:202896576 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTAACACAGCACGACCAATGAAA[G/T]GTGCTAGGAATTATACAGACAGCTT
Phenotype
MIM: 614522 MIM: 603417
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KLHL12 PubMed Links

Gene Details

Gene
KLHL12
Gene Name
kelch like family member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303051.1 Intron NP_001289980.1
NM_001303109.1 Intron NP_001290038.1
NM_021633.3 Intron NP_067646.1
XM_011509835.1 Intron XP_011508137.1
XM_011509836.1 Intron XP_011508138.1
XM_011509837.1 Intron XP_011508139.1
XM_017001995.1 Intron XP_016857484.1
Gene
RABIF
Gene Name
RAB interacting factor
There are no transcripts associated with this gene.

View Full Product Details