Product Details

SNP ID
rs9535527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:50913713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAAGTATTTTAGGAAAGATTGC[A/G]CTTATTATTTATTTCTCCACAGAAA
Phenotype
MIM: 610326
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNASEH2B PubMed Links

Gene Details

Gene
RNASEH2B
Gene Name
ribonuclease H2 subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142279.2 Intron NP_001135751.1
NM_024570.3 Intron NP_078846.2
XM_006719867.3 Intron XP_006719930.1
XM_011535229.1 Intron XP_011533531.1
XM_011535230.2 Intron XP_011533532.1
XM_011535231.2 Intron XP_011533533.1
XM_011535233.2 Intron XP_011533535.1
XM_017020747.1 Intron XP_016876236.1
Gene
RNASEH2B-AS1
Gene Name
RNASEH2B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details