Product Details

SNP ID
rs6502819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4733369 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCCGAGCCTGCCCACAGCTATGC[C/T]TTGGCCTCTCCAGCAGGGTGCTCCT
Phenotype
MIM: 605398 MIM: 612383 MIM: 614312
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXCL16 PubMed Links

Gene Details

Gene
CXCL16
Gene Name
C-X-C motif chemokine ligand 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100812.1 618 Intron NP_001094282.1
NM_022059.3 618 Intron NP_071342.2
Gene
MED11
Gene Name
mediator complex subunit 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001683.3 618 UTR 3 NP_001001683.1
NM_001305000.1 618 UTR 3 NP_001291929.1
Gene
ZMYND15
Gene Name
zinc finger MYND-type containing 15
There are no transcripts associated with this gene.

View Full Product Details