Product Details

SNP ID
rs10406576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13127898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATGTGACCAGGGAGAGTATGAA[A/G]AGCTATGGCTGTCCCATTCCCATTC
Phenotype
MIM: 610672
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NACC1 PubMed Links

Gene Details

Gene
NACC1
Gene Name
nucleus accumbens associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052876.3 Intron NP_443108.1
XM_005259721.3 Intron XP_005259778.1

View Full Product Details