Product Details

SNP ID
rs6012701
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:49633263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGAGGATCTCTGACAAGAGGAC[C/T]TATTATGTCAAAATGAGTATTCTTG
Phenotype
MIM: 604016
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
B4GALT5 PubMed Links

Gene Details

Gene
B4GALT5
Gene Name
beta-1,4-galactosyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004776.3 4410 UTR 3 NP_004767.1

View Full Product Details