Product Details

SNP ID
rs6065993
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46465308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACATACCCATTTTTGGTAAAAT[A/T]AACTGATCCCATGGTTAAGATGATA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MKRN7P PubMed Links

Gene Details

Gene
MKRN7P
Gene Name
makorin ring finger protein 7, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF334
Gene Name
zinc finger protein 334
There are no transcripts associated with this gene.

Gene
ZNF663P
Gene Name
zinc finger protein 663, pseudogene
There are no transcripts associated with this gene.

View Full Product Details